Order Entry
Ireland
ContactUsLinkComponent
Anti-NEAT1 Rabbit Polyclonal Antibody
Anti-NEAT1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7426B
Supplier:  Abgent
undefined
Anti-NEAT1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7426B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vinculin
  • Antigen symbol:
    VCL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    MVCL|Vinculin|CMH15|HEL114|Vinculin |MV|Metavinculin|CMD1W
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: NEAT1 (nuclear paraspeckle assembly transcript 1 (non-protein coding))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse