Anti-MSRA Rabbit Polyclonal Antibody
Catalog # ABGEAP7321A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Methionine sulfoxide reductase A
- Antigen symbol:MSRA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:Peptide methionine sulfoxide reductase|Peptide Met(O) reductase|MSRA|Peptide-methionine (S)-S-oxide reductase|PMSR|Protein-methionine-S-oxide reductase
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MSRA (methionine sulfoxide reductase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse