Anti-SETD8 Rabbit Polyclonal Antibody
Catalog # ABGEAP6557C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:SET Domain Containing 8
- Antigen symbol:SETD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:PR/SET domain-containing protein 07|SET domain-containing protein 8|PRSET7|SETD8|Histone-lysine N-methyltransferase SETD8|SET07|KMT5A|H4-K20-HMTase SETD8|PR/SET07|Lysine N-methyltransferase 5A|SET8|PR-Set7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SETD8 (SET domain containing (lysine methyltransferase) 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human