Order Entry
Ireland
ContactUsLinkComponent
Anti-TBRG1 Rabbit Polyclonal Antibody
Anti-TBRG1 Rabbit Polyclonal Antibody
  ABGEAP6376A
 :  Abgent
undefined
Anti-TBRG1 Rabbit Polyclonal Antibody
  ABGEAP6376A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Transforming Growth Factor Beta Regulator 1
  • Antigen symbol:
    TBRG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    TBRG1|Transforming growth factor beta regulator 1|NIAM|Nuclear interactor of ARF and Mdm2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: TBRG1 (transforming growth factor beta regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human