Anti-PON2 Rabbit Polyclonal Antibody
Catalog # ABGEAP5760C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Paraoxonase 2
- Antigen symbol:PON2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:aromatic esterase 2|serum aryldialkylphosphatase 2|paraoxonase nirs|serum paraoxonase/arylesterase 2|A-esterase 2|PON 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PON2 (paraoxonase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse