Anti-TRIP13 Rabbit Polyclonal Antibody
Catalog # ABGEAP2843A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor Interactor 13
- Antigen symbol:TRIP13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:Human papillomavirus type 16 E1 protein-binding protein|Thyroid hormone receptor interactor 13|TRIP13|16E1-BP|HPV16 E1 protein-binding protein|TRIP-13|Thyroid receptor-interacting protein 13
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TRIP13 (thyroid hormone receptor interactor 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human