Anti-GAS41 (YEATS4/NuBI-1) Rabbit Polyclonal Antibody
ABGEAP1910A
: Abgent
- Antibody type:Primary
- Antigen name:Glioma-amplified sequence-41
- Antigen symbol:GAS41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Antigen synonyms:YEATS4|Glioma-amplified sequence-41|NuBI1|GAS41|YEATS domain-containing protein 4|NuMA binding protein 1|NuBI-1|Gas41
- Sequence:A31
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GAS41 (YEATS4/NuBI-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human