Anti-RPL10A Rabbit Polyclonal Antibody
Catalog # ABGEAP17096A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein L10a
- Antigen symbol:RPL10A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Gene ID:4736
- Antigen synonyms:Csa-19|NEDD6|L10A|CSA19
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:24.831 kDa
- Sequence:LHGNQRKRRKFLETC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RPL10A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 3-31 amino acids from the N-terminal region of human RPL10A.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RPL10A (ribosomal protein L10a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human