Anti-SIGLEC9 Rabbit Polyclonal Antibody
ABGEAP1627B
: Abgent
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 9
- Antigen symbol:SIGLEC9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:CDw329|FOAP-9|siglec-9|OBBP-LIKE|CD329
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SIGLEC9 (sialic acid binding Ig-like lectin 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse