Order Entry
Ireland
ContactUsLinkComponent
Anti-CDH16 Rabbit Polyclonal Antibody
Anti-CDH16 Rabbit Polyclonal Antibody
  ABGEAP13746B
 :  Abgent
undefined
Anti-CDH16 Rabbit Polyclonal Antibody
  ABGEAP13746B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Cadherin 16
  • Antigen symbol:
    CDH16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    CDH16|Kidney-specific Cadherin|CDH 16|Cadherin 16|Ksp-Cadherin|UNQ695/PRO1340
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: CDH16 (cadherin 16, KSP-cadherin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human