Anti-PIASy1 Rabbit Polyclonal Antibody
ABGEAP1250A
: Abgent
- Antibody type:Primary
- Antigen name:Protein inhibitor of activated STAT 4
- Antigen symbol:PIAS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal ( 486–499 aa)
- Cross adsorption:No
- Antigen synonyms:E3 SUMO-protein ligase PIAS4|ZMIZ6|PIAS4|PIAS-gamma|PIASG|PIASY|Piasg|PIASy
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PIASy1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human