Anti-UGT2B15 Rabbit Polyclonal Antibody
Catalog # ABGEAP12375C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:UDP glucuronosyltransferase 2 Family Polypeptide B15
- Antigen symbol:UGT2B15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:ARHGAP47, FKSG15, IDDM21, TAGAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: UGT2B15 (UDP glucuronosyltransferase 2 family, polypeptide B15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human