Anti-ATP6V1B1 Rabbit Polyclonal Antibody
Catalog # ABGEAP12290B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 56/58kDa, V1 Subunit B1
- Antigen symbol:ATP6V1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:ATP6V1B1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ATP6V1B1 (ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human