Anti-TLL2 Rabbit Polyclonal Antibody [clone: 180CT10.2.3]
ABGEAP11319C
: Abgent
- Antibody type:Primary
- Antigen name:Tolloid-like 2
- Antigen symbol:TLL2
- Clonality:Polyclonal
- Clone:180CT10.2.3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:Pyruvate dehydrogenase kinase isoform 2|pyruvate dehydrogenase kinase|PDK2_HUMAN.|mitochondrial|PDHK2|Pyruvate dehydrogenase lipoamide kinase isozyme 2|isozyme 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: TLL2 (tolloid-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human