Order Entry
Ireland
ContactUsLinkComponent
Anti-WDR49 Rabbit Polyclonal Antibody
Anti-WDR49 Rabbit Polyclonal Antibody
Catalog # ABGEAP10872B
Supplier:  Abgent
Anti-WDR49 Rabbit Polyclonal Antibody
Catalog # ABGEAP10872B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    WD Repeat Domain 49
  • Antigen symbol:
    WDR49
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal (27–43 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    D430008P04Rik|TIE|tie-1|JTK14
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: WDR49 (WD repeat domain 49)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human