Anti-INTS10 Rabbit Polyclonal Antibody
Catalog # AVIVARP70587_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Integrator Complex Subunit 10
- Antigen symbol:INTS10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55174
- Antigen synonyms:INT10|C8orf35
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: INTS10 (integrator complex subunit 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human