Anti-GPR141 Rabbit Polyclonal Antibody
AVIVARP68389_P050
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 141
- Antigen symbol:GPR141
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:353345
- Antigen synonyms:PGR13
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GPR141 (G protein-coupled receptor 141)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human