Anti-MBOAT4 Rabbit Polyclonal Antibody
AVIVARP67168_P050
- Antibody type:Primary
- Antigen name:Membrane bound O-acyltransferase domain containing 4
- Antigen symbol:MBOAT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:619373
- Antigen synonyms:FKSG89|GOAT|OACT4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MBOAT4 (membrane bound O-acyltransferase domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: