Anti-SLC35B2 Rabbit Polyclonal Antibody
Catalog # AVIVARP66679_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2
- Antigen symbol:SLC35B2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:347734
- Antigen synonyms:SLL|PAPST1|UGTrel4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC35B2 (solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human