Anti-MS4A7 Rabbit Polyclonal Antibody
Catalog # AVIVARP66405_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-Domains, Subfamily A, Member 7
- Antigen symbol:MS4A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:58475
- Antigen synonyms:MS4A8|CD20L4|CFFM4|4SPAN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MS4A7 (membrane-spanning 4-domains, subfamily A, member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human