Anti-DYNC1LI1 Rabbit Polyclonal Antibody
AVIVARP65751_P050
- Antibody type:Primary
- Antigen name:Dynein cytoplasmic 1, light intermediate chain 1
- Antigen symbol:DYNC1LI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51143
- Antigen synonyms:DNCLI1|LIC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DYNC1LI1 (dynein, cytoplasmic 1, light intermediate chain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human