Anti-NPR3 Rabbit Polyclonal Antibody
AVIVARP64739_P050
- Antibody type:Primary
- Antigen name:Natriuretic peptide receptor 3
- Antigen symbol:NPR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:4883
- Antigen synonyms:NPRC|NPR-C|ANPR-C|GUCY2B|ANP-C|ANPRC|C5orf23
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NPR3 (natriuretic peptide receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human