Anti-TCP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP63618_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:T-Complex 1
- Antigen symbol:TCP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:6950
- Antigen synonyms:CCTa|D6S230E|CCT-alpha|CCT1|TCP-1-alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TCP1 (t-complex 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: