Anti-PGAM1 Rabbit Polyclonal Antibody
AVIVARP61907_P050
- Antibody type:Primary
- Antigen name:Phosphoglycerate Mutase 1
- Antigen symbol:PGAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:5223
- Antigen synonyms:HEL-S-35|PGAMA|PGAM-B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PGAM1 (phosphoglycerate mutase 1 (brain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human