Anti-CLEC4G Rabbit Polyclonal Antibody
Catalog # AVIVARP55883_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:C-type lectin domain family 4, member G
- Antigen symbol:CLEC4G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:339390
- Antigen synonyms:LSECtin|DTTR431|UNQ431|LP2698
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CLEC4G (C-type lectin domain family 4, member G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human