Order Entry
Ireland
ContactUsLinkComponent
Anti-FAM219A Rabbit Polyclonal Antibody
Anti-FAM219A Rabbit Polyclonal Antibody
  AVIVARP55478_P050
undefined
Anti-FAM219A Rabbit Polyclonal Antibody
  AVIVARP55478_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 219, member A
  • Antigen symbol:
    FAM219A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    203259
  • Antigen synonyms:
    bA573M23.5|C9orf25
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: FAM219A (family with sequence similarity 219, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: