Anti-MMADHC Rabbit Polyclonal Antibody
Catalog # AVIVARP55333_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria
- Antigen symbol:MMADHC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:27249
- Antigen synonyms:CL25022|cblD|C2orf25
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MMADHC (methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: