Anti-DENND5A Rabbit Polyclonal Antibody
AVIVARP55199_P050
- Antibody type:Primary
- Antigen name:DENN/MADD domain containing 5A
- Antigen symbol:DENND5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DENND5A (DENN/MADD domain containing 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human