Anti-FBXO8 Rabbit Polyclonal Antibody
AVIVARP54862_P050
- Antibody type:Primary
- Antigen name:F-box protein 8
- Antigen symbol:FBXO8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:26269
- Antigen synonyms:FBX8|FBS|DC10
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: FBXO8 (F-box protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: