Anti-CENPI Rabbit Polyclonal Antibody
Catalog # AVIVARP54805_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Centromere Protein I
- Antigen symbol:CENPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2491
- Antigen synonyms:CENP-I|FSHPRH1|LRPR1|Mis6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CENPI (centromere protein I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human