Anti-ZC3H14 Rabbit Polyclonal Antibody
Catalog # AVIVARP53850_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger CCCH-type containing 14
- Antigen symbol:ZC3H14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:79882
- Antigen synonyms:MSUT-2|UKp68|SUT2|NY-REN-37
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZC3H14 (zinc finger CCCH-type containing 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: