Anti-NKAIN4 Rabbit Polyclonal Antibody
AVIVARP53118_P050
- Antibody type:Primary
- Antigen name:Na+/K+ transporting ATPase interacting 4
- Antigen symbol:NKAIN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:128414
- Antigen synonyms:C20orf58|FAM77A|bA261N11.2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NKAIN4 (Na+/K+ transporting ATPase interacting 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: