Anti-CCSAP Rabbit Polyclonal Antibody
Catalog # AVIVARP52834_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Centriole, cilia and spindle-associated Protein
- Antigen symbol:CCSAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:126731
- Antigen synonyms:CSAP|C1orf96
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CCSAP (centriole, cilia and spindle-associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human