Anti-ARID5A Rabbit Polyclonal Antibody
Catalog # AVIVARP50912_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:AT rich interactive domain 5A (MRF1-like)
- Antigen symbol:ARID5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10865
- Antigen synonyms:RP11-363D14|MRF-1|MRF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ARID5A (AT rich interactive domain 5A (MRF1-like))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: