Anti-NRSN2 Rabbit Polyclonal Antibody
AVIVARP49914_P050
- Antibody type:Primary
- Antigen name:Neurensin 2
- Antigen symbol:NRSN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:80023
- Antigen synonyms:dJ1103G7.6|C20orf98
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NRSN2 (neurensin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: