Anti-PLXDC1 Rabbit Polyclonal Antibody
AVIVARP49500_P050
- Antibody type:Primary
- Antigen name:Plexin domain containing 1
- Antigen symbol:PLXDC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57125
- Antigen synonyms:TEM3|TEM7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PLXDC1 (plexin domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: