Anti-YIPF1 Rabbit Polyclonal Antibody
AVIVARP49421_P050
- Antibody type:Primary
- Antigen name:Yip1 domain family, member 1
- Antigen symbol:YIPF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:54432
- Antigen synonyms:FinGER1|DJ167A19.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: YIPF1 (Yip1 domain family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: