Anti-ESCO1 Rabbit Polyclonal Antibody
AVIVARP48979_P050
- Antibody type:Primary
- Antigen name:Establishment of sister chromatid cohesion N-acetyltransferase 1
- Antigen symbol:ESCO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:114799
- Antigen synonyms:A930014I12Rik|CTF|ESO1|EFO1|ECO1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ESCO1 (establishment of sister chromatid cohesion N-acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: