Anti-NR2F1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48000_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor subfamily 2, group F, member 1
- Antigen symbol:NR2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Pig,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:7025
- Antigen synonyms:EAR-3|EAR3|BBSOAS|ERBAL3|TCFCOUP1|SVP44|TFCOUP1|BBOAS|NR2F2|COUP-TFI
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NR2F1 (nuclear receptor subfamily 2, group F, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat