Anti-ATP6V0C Rabbit Polyclonal Antibody
AVIVARP45161_P050
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c
- Antigen symbol:ATP6V0C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:527
- Antigen synonyms:Vma3|ATPL|VATL|ATP6C|VPPC|ATP6L
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ATP6V0C (ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: