Anti-SLC29A2 Rabbit Polyclonal Antibody
Catalog # AVIVARP43822_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 29 (equilibrative nucleoside transporter), member 2
- Antigen symbol:Solute carrier family 29, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3177
- Antigen synonyms:HNP36|ENT2|DER12
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC29A2 (solute carrier family 29 (equilibrative nucleoside transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human