Anti-GZMH Rabbit Polyclonal Antibody
Catalog # AVIVARP42686_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Granzyme H
- Antigen symbol:GZMH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2999
- Antigen synonyms:CGL-2|CCP-X|CTLA1|CTSGL2|CSP-C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GZMH (granzyme H (cathepsin G-like 2, protein h-CCPX))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human