Anti-ZKSCAN8 Rabbit Polyclonal Antibody
AVIVARP37750_P050
- Antibody type:Primary
- Antigen name:zinc finger with KRAB and SCAN domains 8
- Antigen symbol:ZKSCAN8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7745
- Antigen synonyms:ZNF192|ZSCAN40|LD5-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZKSCAN8 (zinc finger with KRAB and SCAN domains 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: