Anti-ETV2 Rabbit Polyclonal Antibody
Catalog # AVIVARP36850_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ETS variant 2
- Antigen symbol:ETV2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:14008
- Antigen synonyms:Etsrp71
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ETV2 (ets variant 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: