Anti-ZNF248 Rabbit Polyclonal Antibody
AVIVARP35992_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 248
- Antigen symbol:ZNF248
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57209
- Antigen synonyms:bA162G10.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF248 (zinc finger protein 248)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human