Order Entry
Ireland
ContactUsLinkComponent
Anti-VEZF1 Rabbit Polyclonal Antibody
Anti-VEZF1 Rabbit Polyclonal Antibody
  AVIVARP35830_P050
Anti-VEZF1 Rabbit Polyclonal Antibody
  AVIVARP35830_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Vascular endothelial zinc finger 1
  • Antigen symbol:
    VEZF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    7716
  • Antigen synonyms:
    ZNF161|DB1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: VEZF1 (vascular endothelial zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: