Anti-PCSK1 Rabbit Polyclonal Antibody
Catalog # AVIVARP33848_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Proprotein convertase subtilisin/kexin type 1
- Antigen symbol:PCSK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:5122
- Antigen synonyms:SPC3|PC3|BMIQ12|PC1|NEC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PCSK1 (proprotein convertase subtilisin/kexin type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: