Anti-TSFM Rabbit Polyclonal Antibody
Catalog # AVIVARP31455_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ts translation elongation factor, mitochondrial
- Antigen symbol:TSFM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:10102
- Antigen synonyms:EFTSMT|EFTS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TSFM (Ts translation elongation factor, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: