Laboratory coats, DuPont style, TPR250
Lab coat made of Tyvek®. Suitable for protection against a variety of liquid or solid chemicals, as well as fine particulates and fibres and biological hazards.
- With press stud front
- Without pockets
- With collar
- Available with or without elasticated cuffs
Colour: White
- Catalog No:
- 113-1765
- 113-1751
- 113-1752
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Garment application:
- Laboratory
- Laboratory
- Laboratory
Frequently Bought Together