Water sampling bottles, glass, narrow neck, Boston type, round
These bottles, made of Type III soda-lime glas, clear or amber, with cap, are recommended for collecting water samples for variety of organic and inorganic target analytes where light protection is not required.
- With PTFE-lined white PP closed top cap (assembled on bottles)
- Certified to meet EPA performance based specifications for organics, pesticides and metal anlyses or unprocessed and readily assembled for own cleaning procedure
- Each jar individually barcoded with lot number and unique container number
: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
: Carton sealed with security tape providing tamper evidence. The chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping.
- Catalog No:
- EPSC349-0500
- EPSC329-0250
- EPSC129-0500
- EPSC229-0500
- EPSC349-0125
- EPSC129-1000
- EPSC229-1000
- EPSC329-1000
- EPSC129-0125
- EPSC229-0125
- EPSC129-0250
- EPSC229-0250
- EPSC329-0500
- EPSC329-0125
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together